CAS RN 899431-18-6
IUPAC Nombre:
3-{[5-(2,3-dichlorophenyl)-1H-1,2,3,4-tetrazol-1-yl]methyl}pyridine hydrochloride
Sinónimos:
3-{[5-(2,3-dichlorophenyl)-1,2,3,4-tetrazol-1-yl]methyl}pyridine hydrochloride
Peso molecular (g/mol):
342.61
Fórmula molecular:
C13H10Cl3N5
InChi Key:
MBTJFFMIPPMRGR-UHFFFAOYSA-N
SMILES:
Cl.ClC1=CC=CC(C2=NN=NN2CC2=CN=CC=C2)=C1Cl
1
–
2
de
2
resultados
1
Tocris Bioscience™ A 438079 hydrochloride
CAS: 899431-18-6 Fórmula molecular: C13H10Cl3N5
2
A 438079 hydrochloride, MedChemExpress
MedChemExpress A 438079 (hydrochloride) is a potent, and selective P2X7 receptor antagonist with pIC50 of 6.9.