CAS RN 890643-16-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
DC-05, MedChemExpress
MedChemExpress DC-05 is a DNA methyltransferase 1 (DNMT1) inhibitor, with an IC50 and a Kd of 10.3 μM and 1.09 μM, respectively.