CAS RN 88898-17-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
MedChemExpress Atrial Natriuretic Peptide (ANP) (1-28), rat
MedChemExpress Atrial Natriuretic Peptide (ANP) (1-28), rat is a major circulating form of ANP in rats, potently inhibits Angiotensin II (Ang II)-stimulated endothelin-1 secretion in a concentration-dependent manner.
2
MedChemExpress Atrial Natriuretic Peptide (ANP) (1-28), rat
MedChemExpress Atrial Natriuretic Peptide (ANP) (1-28), rat is a major circulating form of ANP in rats, potently inhibits Angiotensin II (Ang II)-stimulated endothelin-1 secretion in a concentration-dependent manner.