CAS RN 886047-22-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
1
Tocris Bioscience™ LY 2119620
Positive allosteric modulator of M2 and M4 receptors
2
Tocris Bioscience™ LY 2119620
Positive allosteric modulator of M2 and M4 receptors
3
LY2119620, MedChemExpress
MedChemExpress LY2119620 is a high-affinity muscarinic M2/M4 receptor agonist.