CAS RN 872365-14-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Fevipiprant, MedChemExpress
MedChemExpress Fevipiprant (QAW039; NVP-QAW039) is a selective, potent, reversible competitive CRTh2 antagonist with an in vitro dissociation constant KD value of 1.1nM at the CRTh2 receptor and an IC50 value of 0.44 nM for inhibition of PGD2-induced eosinophil shape change in human whole blood.