CAS RN 867160-71-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
2
Tocris Bioscience™ Linsitinib
Potent and selective dual inhibitor of the IGF-1 receptor and insulin receptor
3
Linsitinib, MedChemExpress
MedChemExpress Linsitinib (OSI-906) is a potent, selective and orally bioavailable dual inhibitor of the IGF-1 receptor and insulin receptor (IR) with IC50s of 35 and 75 nM, respectively.