CAS RN 858128-57-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
SPB, MedChemExpress
MedChemExpress SPB is a drug-linker conjugate for ADC with potent anti-inflammatory activity by using Xanthotoxol, linked via the ADC linker.