CAS RN 856095-68-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
OBA-09, MedChemExpress
MedChemExpress OBA-09, a simple ester of pyruvate and salicylic acid, is potent multi-modal neuroprotectant. OBA-09 has anti-oxidative and anti-inflammatory effects.