CAS RN 853400-76-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
1
Trofinetida (>90%), TRC
CAS: 853400-76-7 Fórmula molecular: C13H21N3O6 Peso molecular (g/mol): 315.32 Sinónimo: Glycyl-2-methyl-L-prolyl-L-glutamic Acid,Glycyl-L-2-methylprolyl-L-glutamic Acid,NNZ 2566 Nombre IUPAC: (2S)-2-[[(2S)-1-(2-aminoacetil)-2-metilpirrolidina-2-carbonil]amino]pentanedioico SMILES: C[C@]1(CCCN1C(=O)CN)C(=O)N[C@@H](CCC(=O)O)C(=O)O
2
Tocris Bioscience™ Trofinetide
IGF-1 synthetic analogue
3
Trofinetide, MedChemExpress
MedChemExpress Trofinetide (NNZ-2566), a synthetic analogue of the endogenous N-terminus tripeptide, Glycine-Proline-Glutamate (GPE), has been shown to be neuroprotective in animal models of brain injury.