CAS RN 852329-66-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Dutogliptin, MedChemExpress
MedChemExpress Dutogliptin (PHX-1149 free base) is an orally available, potent, and selective dipeptidyl peptidase-4 (DPP4) inhibitor for the treatment of type 2 diabetes mellitus.