CAS RN 851983-85-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
5
de
5
resultados
2
Tocris Bioscience™ Galeterone
CYP17 inhibitor; also androgen receptor antagonist
3
Galeterone, MedChemExpress
MedChemExpress Galeterone (TOK-001) is a multifunctional antiandrogen and CYP17 inhibitor (IC50=47 nM) in castration resistant prostate cancer (CRPC).
4
5