CAS RN 850717-64-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
BX517, MedChemExpress
MedChemExpress BX517 is a potent and selective inhibitor of PDK1 with IC50 of 6 nM.