CAS RN 850173-95-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
ADL-5859, MedChemExpress
MedChemExpress ADL5859 is a (delta)-opioid receptor agonist with Ki of 0.8 nM, selectivity against opioid receptor (kappa), (mu), and weak inhibitory activity at the hERG channel.