CAS RN 849550-05-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Cevipabulin, MedChemExpress
MedChemExpress Cevipabulin (TTI-237) is an oral, microtubule-active antitumor compound and inhibits the binding of [3H] vinblastine to tubulin, with an IC50 of 18-40 nM for cytotoxicity in human tumor cell line.