CAS RN 840529-13-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Helioxanthin 8-1, MedChemExpress
MedChemExpress Helioxanthin 8-1 is an analogue of helioxanthin, exhibites significant in vitro anti-HBV/HCV/HSV-1/HIV activity with EC50 of >5/10/1.4/15 uM.