CAS RN 839707-37-8
IUPAC Nombre:
N-(3-{7-[(1,3-dimethyl-1H-pyrazol-5-yl)amino]-1-methyl-2-oxo-1H,2H,3H,4H-[1,3]diazino[4,5-d]pyrimidin-3-yl}-4-methylphenyl)-3-(trifluoromethyl)benzamide
Sinónimos:
N-(3-{7-[(2,5-dimethylpyrazol-3-yl)amino]-1-methyl-2-oxo-4H-[1,3]diazino[4,5-d]pyrimidin-3-yl}-4-methylphenyl)-3-(trifluoromethyl)benzamide
Peso molecular (g/mol):
550.55
Fórmula molecular:
C27H25F3N8O2
InChi Key:
NBZFRTJWEIHFPF-UHFFFAOYSA-N
SMILES:
CN1N=C(C)C=C1NC1=NC=C2CN(C(=O)N(C)C2=N1)C1=C(C)C=CC(NC(=O)C2=CC(=CC=C2)C(F)(F)F)=C1
1
–
2
de
2
resultados
1
Pluripotin, MedChemExpress
MedChemExpress Pluripotin is a dual inhibitor of ERK1 and RasGAP with KDs of 98 nM and 212 nM, respectively. Pluripotin also inhibits RSK1, RSK2, RSK3, and RSK4 with IC50s of 0.5, 2.5, 3.3, and 10.0 μM, respectively.
2