CAS RN 83625-35-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Amebucort, MedChemExpress
MedChemExpress Amebucort is a synthetic glucocorticoid corticosteroid, may used for the research of inflammatory disorders.