CAS RN 82692-93-1
IUPAC Nombre:
sodium 3-[ethyl(3-methylphenyl)amino]-2-hydroxypropane-1-sulfonate
Sinónimos:
sodium 3-[ethyl(3-methylphenyl)amino]-2-hydroxypropane-1-sulfonate
Peso molecular (g/mol):
295.33
Fórmula molecular:
C12H18NNaO4S
InChi Key:
IRQRBVOQGUPTLG-UHFFFAOYNA-M
SMILES:
[Na+].CCN(CC(O)CS([O-])(=O)=O)C1=CC=CC(C)=C1
1
–
1
de
1
resultados
1
TOOS, MedChemExpress
MedChemExpress TOOS, a Trinder's reagent, is a novel highly water-soluble aniline derivative; are widely used in diagnostic tests and biochemical tests.