CAS RN 81624-55-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
NSC348884, MedChemExpress
MedChemExpress NSC348884 is a nucleophosmin inhibitor disrupts oligomer formation and induces apoptosis, inhibits cell proliferation at an IC50 of 1.7-4.0 μM in distinct cancer cell lines.