CAS RN 81382-51-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Pentiapine, MedChemExpress
MedChemExpress Pentiapine (CGS 10746) is a dopamine release inhibitor without binding to synaptic dopamine receptor sites.