CAS RN 81117-35-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
N-(n-Nonil)desoxinojirimicina, TRC
CAS: 81117-35-3 Fórmula molecular: C15 H31 N O4 Peso molecular (g/mol): 289.41 Sinónimo: N-Nonyldeoxynojirimycin Nombre IUPAC: (2R,3R,4R,5S)-2-(hidroximetil)-1-nonilpiperidina-3,4,5-triol SMILES: CCCCCCCCCN1C[C@H](O)[C@@H](O)[C@H](O)[C@H]1CO
2
MedChemExpress N-Nonyldeoxynojirimycin
MedChemExpress N-Nonyldeoxynojirimycin (NN-DNJ) is a potent inhibitor of alpha-glucosidase and alpha-1,6-glucosidase (IC50s, 0.42, 8.4 μM, respectively), inhibits glycogen breakdown.