CAS RN 79955-27-4
IUPAC Nombre:
3',6'-bis(acetyloxy)-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthene]-5-carboxylic acid
Sinónimos:
3',6'-bis(acetyloxy)-3-oxospiro[2-benzofuran-1,9'-xanthene]-5-carboxylic acid
Peso molecular (g/mol):
460.39
Fórmula molecular:
C25H16O9
InChi Key:
WPUZGNPQMIWOHE-UHFFFAOYSA-N
SMILES:
CC(=O)OC1=CC=C2C(OC3=CC(OC(C)=O)=CC=C3C22OC(=O)C3=CC(=CC=C23)C(O)=O)=C1
1
–
1
de
1
resultados
1
MedChemExpress 5-CFDA
MedChemExpress 5-CFDAis membrane-permeant and can be loaded into cells via incubation; hydrolyzed by intracellular esterases to 5-carboxyfluorescein; used for labeling human intervertebral disk cells in vitro for fluorescence microscopy.