CAS RN 79714-31-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Risarestat, MedChemExpress
MedChemExpress Risarestat (CT-112), an aldose reductase inhibitor, is developed for the treatment of diabetic complications.