CAS RN 79517-01-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
1
Octreotide Acetate, TRC
CAS: 79517-01-4
2
MedChemExpress Octreotide acetate
MedChemExpress Octreotide acetate, a long-acting synthetic analog of native somatostatin, inhibits growth hormone, glucagon, and insulin more potently.
3
MedChemExpress Octreotide acetate
MedChemExpress Octreotide acetate, a long-acting synthetic analog of native somatostatin, inhibits growth hormone, glucagon, and insulin more potently.