CAS RN 77529-42-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Glycolic acid oxidase inhibitor 1, MedChemExpress
MedChemExpress Glycolic acid oxidase inhibitor 1 is a glycolate oxidase inhibitor, extracted from patent EP0021228A1, in Table IV.