CAS RN 755038-02-9
IUPAC Nombre:
4-[(8-cyclopentyl-7-ethyl-5-methyl-6-oxo-5,6,7,8-tetrahydropteridin-2-yl)amino]-3-methoxy-N-(1-methylpiperidin-4-yl)benzamide
Sinónimos:
4-[(8-cyclopentyl-7-ethyl-5-methyl-6-oxo-7H-pteridin-2-yl)amino]-3-methoxy-N-(1-methylpiperidin-4-yl)benzamide
Peso molecular (g/mol):
521.67
Fórmula molecular:
C28H39N7O3
InChi Key:
XQVVPGYIWAGRNI-UHFFFAOYNA-N
SMILES:
CCC1N(C2CCCC2)C2=NC(NC3=C(OC)C=C(C=C3)C(=O)NC3CCN(C)CC3)=NC=C2N(C)C1=O
1
–
2
de
2
resultados
2
BI 2536, MedChemExpress
MedChemExpress BI 2536 is a dual PLK1 and BRD4 inhibitor with IC50s of 0.83 and 25 nM, respectively. BI-2536 suppresses IFNB (encoding IFN-β) gene transcription.