CAS RN 7497-07-6
IUPAC Nombre:
3,4-dichloro-N-(pentan-2-yl)benzamide
Sinónimos:
3,4-dichloro-N-(pentan-2-yl)benzamide
Peso molecular (g/mol):
260.16
Fórmula molecular:
C12H15Cl2NO
InChi Key:
ARDYECYBETXQFD-UHFFFAOYNA-N
SMILES:
CCCC(C)NC(=O)C1=CC=C(Cl)C(Cl)=C1
1
–
1
de
1
resultados
1
NSC 405020, MedChemExpress
MedChemExpress NSC 405020 is a novel small molecule inhibitor of MT1-MMP that specifically targets PEX domain rather than the catalytic domain of MT1-MMP with IC50 >100 μM and does not inhibit the catalytic activity of MT1-MMP or MMP-2.