CAS RN 749234-11-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
DMAT, MedChemExpress
MedChemExpress DMAT is a potent and specific CK2 inhibitor with an IC50 value of 130 nM.