CAS RN 727664-91-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
MLS0315771, MedChemExpress
MedChemExpress MLS0315771 is a potent and biologically active competitive phosphomannose isomerase (MPI) inhibitor, with an IC50 ∼1 μM and a Ki of 1.4 μM.