CAS RN 72093-21-1
IUPAC Nombre:
(2S)-N-[(1S)-1-{[(1S)-5-amino-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-5-amino-1-{[(1S)-5-amino-1-{[(1S)-1-{[(1S)-1-carbamoyl-3-methylbutyl]carbamoyl}-2-methylbutyl]carbamoyl}pentyl]carbamoyl}pentyl]carbamoyl}ethyl]carbamoyl}-3-methylbutyl]carbamoyl}ethyl]carbamoyl}ethyl]carbamoyl}-3-methylbutyl]carbamoyl}ethyl]carbamoyl}pentyl]carbamoyl}-3-methylbutyl]-2-[(2S,3S)-2-amino-3-methylpentanamido]butanediamide
Sinónimos:
(2S)-N-[(1S)-1-{[(1S)-5-amino-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-5-amino-1-{[(1S)-5-amino-1-{[(1S)-1-{[(1S)-1-carbamoyl-3-methylbutyl]carbamoyl}-2-methylbutyl]carbamoyl}pentyl]carbamoyl}pentyl]carbamoyl}ethyl]carbamoyl}-3-methylbutyl]carbamoyl}ethyl]carbamoyl}ethyl]carbamoyl}-3-methylbutyl]carbamoyl}ethyl]carbamoyl}pentyl]carbamoyl}-3-methylbutyl]-2-[(2S,3S)-2-amino-3-methylpentanamido]succinamide
Peso molecular (g/mol):
1478.94
Fórmula molecular:
C70H131N19O15
InChi Key:
MASXKPLGZRMBJF-HDSAQESISA-N
SMILES:
CC[C@H](C)[C@H](N)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](C)C(=O)N[C@@H](C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](C)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](C(C)CC)C(=O)N[C@@H](CC(C)C)C(N)=O
1
–
4
de
4
resultados
1
MedChemExpress Mastoparan
MedChemExpress Mastoparan, a tetradecapeptide which is a component of wasp venom, stimulates release of prolactin from cultured rat anterior pituitary cells.
2
MedChemExpress Mastoparan
MedChemExpress Mastoparan, a tetradecapeptide which is a component of wasp venom, stimulates release of prolactin from cultured rat anterior pituitary cells.
3
MedChemExpress Mastoparan
MedChemExpress Mastoparan, a tetradecapeptide which is a component of wasp venom, stimulates release of prolactin from cultured rat anterior pituitary cells.
4
MedChemExpress Mastoparan
MedChemExpress Mastoparan, a tetradecapeptide which is a component of wasp venom, stimulates release of prolactin from cultured rat anterior pituitary cells.