CAS RN 7000-27-3
IUPAC Nombre:
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
Sinónimos:
O1-methyl-glucose β-methylglucoside β-d-methylglucopyranoside M0779_sigma MGL methyl hexopyranoside β-methyl-d-glucoside methyl β-d-glucoside
Peso molecular (g/mol):
194.18
Fórmula molecular:
C7H14O6
InChi Key:
HOVAGTYPODGVJG-XUUWZHRGSA-N
SMILES:
CO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O