CAS RN 697797-51-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
UAMC00039 dihydrochloride, MedChemExpress
MedChemExpress UAMC00039 dihydrochloride is a potent, reversible and competitive dipeptidyl peptidase II inhibitor with an IC50 of 0.48 nM.