CAS RN 69552-46-1
IUPAC Nombre:
5-[(2E,3aS,4R,5R,6aS)-5-hydroxy-4-[(1E,3S)-3-hydroxyoct-1-en-1-yl]-octahydropentalen-2-ylidene]pentanoic acid
Sinónimos:
5-[(2E,3aS,4R,5R,6aS)-5-hydroxy-4-[(1E,3S)-3-hydroxyoct-1-en-1-yl]-hexahydro-1H-pentalen-2-ylidene]pentanoic acid
Peso molecular (g/mol):
350.5
Fórmula molecular:
C21H34O4
InChi Key:
XZFRIPGNUQRGPI-WBQKLGIQSA-N
SMILES:
CCCCC[C@H](O)\C=C\[C@H]1[C@H](O)C[C@@H]2C\C(C[C@H]12)=C/CCCC(O)=O
1
–
1
de
1
resultados
1
Carbacyclin, MedChemExpress
MedChemExpress Carbacyclin is a PGI2 analogue, acts as a prostacyclin (PGI2) receptor agonist and vasodilator, and potently inhibits platelet aggregation.