CAS RN 680-31-9
IUPAC Nombre:
[bis(dimethylamino)phosphoryl]dimethylamine
Sinónimos:
hexamethylphosphoramide hexamethylphosphorsaeuretriamid hexamethylphosphoric-triamide hmpta hempa phosphoric hexamethyltriamide hexamethylphosphoramid HMPA HMPT phosphoric tris(dimethylamide)
Peso molecular (g/mol):
179.2
Fórmula molecular:
C6H18N3OP
InChi Key:
GNOIPBMMFNIUFM-UHFFFAOYSA-N
SMILES:
CN(C)P(=O)(N(C)C)N(C)C
1
–
3
de
3
resultados
2
Hexametiltriamidofosfato, TRC
CAS: 680-31-9 Fórmula molecular: C6 H18 N3 O P Peso molecular (g/mol): 179.2 Sinónimo: HMPA,Hexamethylphosphoric triamide Nombre IUPAC: N-[bis(dimetilamino)fosforil]-N-metilmetanamina SMILES: CN(C)P(=O)(N(C)C)N(C)C