CAS RN 63638-91-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Brofaromine, MedChemExpress
MedChemExpress Brofaromine (CGP 11305A) is a monoamine oxidase (MAO) inhibitor with IC50 of 0.2 μM for MAO-A.