CAS RN 63550-99-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Rebaudioside C, TRC
CAS: 63550-99-2 Fórmula molecular: C44 H70 O22 Peso molecular (g/mol): 951.01 Sinónimo: Kaur-16-en-18-oic acid, 13-[(O-6-deoxy-α-L-mannopyranosyl-(1→2)-O-[β-D-glucopyranosyl-(1→3)]-β-D-glucopyranosyl)oxy]-, β-D-glucopyranosyl ester, (4α)-,Dulcoside B,Rebaudioside C SMILES: C[C@@H]1O[C@@H](O[C@H]2[C@H](O[C@]34CC[C@H]5[C@]6(C)CCC[C@](C)([C@H]6CC[C@@]5(CC3=C)C4)C(=O)O[C@@H]7O[C@H](CO)[C@@H](O)[C@H](O)[C@H]7O)O[C@H](CO)[C@@H](O)[C@@H]2O[C@@H]8O[C@H](CO)[C@@H](O)[C@H](O)[C@H]8O)[C@H](O)[C@H](O)[C@H]1O
2
Rebaudioside C, MedChemExpress
MedChemExpress Rebaudioside C(Dulcoside B) is used as natural sweeteners to diabetics and others on carbohydrate-controlled diets.