CAS RN 635323-95-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
F-15599, MedChemExpress
MedChemExpress F-15599 is a highly selective G-protein biased 5-HT1A receptor agonist, with Ki of 3.4 nM.