CAS RN 614-35-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
p-Synephrine, MedChemExpress
MedChemExpress p-Synephrine is an organic compound, found in multiple biofluids, such as urine and blood.