CAS RN 61036-62-2
IUPAC Nombre:
(1S,2R,19R,22R,34S,37R,40R,52R)-22-amino-63,65-dichloro-64-{[(2S,3R,4R,5S,6R)-3-decanamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-2-{[(2R,3R,4R,5S,6R)-3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-26,31,44,49-tetrahydroxy-21,35,38,54,56,59-hexaoxo-47-{[(2R,3S,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-7,13,28-trioxa-20,36,39,53,55,58-hexaazaundecacyclo[38.14.2.2³,⁶.2¹⁴,¹⁷.2¹⁹,³⁴.1⁸,¹².1²³,²⁷.1²⁹,³³.1⁴¹,⁴⁵.0¹⁰,³⁷.0⁴⁶,⁵¹]hexahexaconta-3,5,8,10,12(64),14,16,23,25,27(61),29,31,33(60),41,43,45(57),46,48,50,62,65-henicosaene-52-carboxylic acid
Sinónimos:
teicoplanin
Peso molecular (g/mol):
1879.67
Fórmula molecular:
C88H97Cl2N9O33
InChi Key:
BJNLLBUOHPVGFT-QRZIFLFXSA-N
SMILES:
CCCCCCCCCC(=O)N[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]1OC1=C2OC3=CC=C(C[C@H]4NC(=O)[C@H](N)C5=CC=C(O)C(OC6=CC(O)=CC(=C6)[C@H](NC4=O)C(=O)N[C@@H]4C(C=C1OC1=CC=C(C=C1Cl)[C@@H](O[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1NC(C)=O)[C@@H]1NC(=O)[C@H](NC4=O)C4=CC=C(O)C(=C4)C4=C(O[C@H]6O[C@H](CO)[C@@H](O)[C@H](O)[C@@H]6O)C=C(O)C=C4[C@@H](NC1=O)C(O)=O)=C2)=C5)C=C3Cl
1
–
3
de
3
resultados
2
Teicoplanin, MedChemExpress
MedChemExpress Teicoplanin is a glycopeptide antibiotic used in the prophylaxis and treatment of serious infections caused by Gram-positive bacteria, including Methicillin-resistant Staphylococcus aureus and Enterococcus faecalis.
3
Melford Teicoplanin
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.