CAS RN 606-17-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
2
Iodipamide, MedChemExpress
MedChemExpress Iodipamide is a tri-iodinated benzoate derivative and ionic dimeric contrast agent used in diagnostic imaging.