CAS RN 6055-72-7
IUPAC Nombre:
7H-purin-6-amine hydrate hydrochloride
Sinónimos:
adenine hydrate hydrochloride
Peso molecular (g/mol):
189.6
Fórmula molecular:
C5H8ClN5O
InChi Key:
MYRDTAUFFBYTHA-UHFFFAOYSA-N
SMILES:
O.Cl.NC1=C2NC=NC2=NC=N1
1
–
3
de
3
resultados
2
Clorhidrato de adenina, ∼99%, MP Biomedicals™
CAS: 6055-72-7 Fórmula molecular: C5H8ClN5O Peso molecular (g/mol): 189.60 Número MDL: MFCD11046306 Clave InChI: MYRDTAUFFBYTHA-UHFFFAOYSA-N Sinónimo: 7h-purin-6-amine hydrochloride hydrate,adenine hydrochloride hydrate,adenine hydrochloride hemihydrate,1h-purin-6-amine monohydrochloride hydrate 2:1,ade hydrate hydrochloride,purine-6-ylamine, chloride, hydrate,c5h5n5.hcl.h2o,adenine hydrochloride,adeninehydrochloridehemihydrate,vitamin b4 hydrate hydrochloride PubChem CID: 24211900 Nombre IUPAC: hidrocloruro de 7H-purin-6-amina hidratada SMILES: O.Cl.NC1=C2NC=NC2=NC=N1