CAS RN 601514-19-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
1
Tocris Bioscience™ TWS 119
Potent GSK3 inhibitor; induces neuronal and CD8(+) T cell differentiation
2
3
TWS119, MedChemExpress
MedChemExpress TWS119 is a specific inhibitor of GSK-3β, with an IC50 of 30 nM, and activates the wnt/β-catenin pathway.