CAS RN 59989-18-3
IUPAC Nombre:
5-ethynyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Sinónimos:
5-ethynyluracil
Peso molecular (g/mol):
136.11
Fórmula molecular:
C6H4N2O2
InChi Key:
JOZGNYDSEBIJDH-UHFFFAOYSA-N
SMILES:
O=C1NC=C(C#C)C(=O)N1
1
–
1
de
1
resultados
1
Eniluracil, MedChemExpress
MedChemExpress Eniluracil (5-Ethynyluracil), a uracil analogue and a mechanism-based irreversible inhibitor of dihydropyrimidine dehydrogenase (DPD), increases the oral bioavailability of 5-fluorouracil (5-FU) to 100%, facilitating uniform absorption and predictable toxicity.