CAS RN 585543-15-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Losmapimod, MedChemExpress
MedChemExpress Losmapimod (GSK-AHAB) is a selective, potent, and orally active p38 MAPK inhibitor with pKis of 8.1 and 7.6 for p38α and p38β, respectively.