CAS RN 57966-42-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
4
de
4
resultados
1
MedChemExpress Proctolin
MedChemExpress Proctolin is an endogenous pentapeptide that acts as an excitatory neuromodulator.
2
MedChemExpress Proctolin
MedChemExpress Proctolin is an endogenous pentapeptide that acts as an excitatory neuromodulator.
3
MedChemExpress Proctolin
MedChemExpress Proctolin is an endogenous pentapeptide that acts as an excitatory neuromodulator.
4
MedChemExpress Proctolin
MedChemExpress Proctolin is an endogenous pentapeptide that acts as an excitatory neuromodulator.