CAS RN 575474-82-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
R112, MedChemExpress
MedChemExpress R112 is an ATP-competitive inhibitor of Syk kinase with a Ki of 96 nM. R112 inhibits Syk kinase activity with an IC50 of 226 nM.