CAS RN 5749-67-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Carbasalato de Calcio, TRC
CAS: 5749-67-7 Fórmula molecular: 2(C9H7O4) . CH4N2O . Ca Peso molecular (g/mol): 458.4 Sinónimo: Calpirinsan,Calcium acetyl salicylate carbamide,Iromin,Omegin,Solupsan,SoluspanUrea calcium acetylsalicylate,Ursinus,Salicylic acid acetate, calcium salt, compd. with urea,Acetylsalicylic acid calcium salt complex with urea Nombre IUPAC: calcio; 2-acetiloxibenzoato; urea SMILES: [Ca+2].CC(=O)Oc1ccccc1C(=O)[O-].CC(=O)Oc2ccccc2C(=O)[O-].NC(=O)N
2
Carbasalate calcium, MedChemExpress
MedChemExpress Carbasalate calcium is an anti-inflammatory, antipyretic, and analgesic agent.