CAS RN 57420-46-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Barlerin, MedChemExpress
MedChemExpress Barlerin (8-O-Acetyl shanzhiside methyl ester) is an iridoid glucoside isolated from the leaves of Lamiophlomis rotata Kudo, a Chinese folk medicinal plant in Xi-zang. Barlerin (8-O-Acetyl shanzhiside methyl ester) could inhibt NF-κB.