CAS RN 566939-85-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Orteronel, MedChemExpress
MedChemExpress Orteronel (TAK-700) is a highly selective inhibitor of human 17,20-lyase (CYP17) with IC50 of 38 nM, and exhibits >1000-fold selectivity over other CYPs such as 11-hydroxylase and CYP3A4.