CAS RN 566203-88-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
AZD-2098, TRC
CAS: 566203-88-1 Fórmula molecular: C11H9Cl2N3O3S Peso molecular (g/mol): 334.18 SMILES: COc1nccnc1NS(=O)(=O)c2cccc(Cl)c2Cl
2
AZD2098, MedChemExpress
MedChemExpress AZD2098 is a potent and selective CC-chemokine receptor 4 (CCR4) inhibitor with pIC50s of 7.8, 8.0, 8.0 and 7.6 for human, rat, mouse and dog respectively, used for asthma research.